Learn More
Moldex™ 8000 Particulate Filters
Click to view available options
Filter Type:
P2RD
P3RD
Unit Size:
Pack of 8
Description
Moldex 8000 Series P2 R D filters (8070). P2 R D particulate filters for the 8000 Series half mask. For protection against harmful and carcinogenic dusts, fumes and aerosols based on water and oil. 10 x WEL.
Moldex 8000 Series P3 R D filters (8080). P3 R D particulate filters for the 8000 Series half mask. For protection against harmful and carcinogenic dusts, fumes and aerosols including radioactive particulates as well as airborne biological substances and enzymes. 20 x WEL.
Moldex 8000 Series P3 R D filters (8080). P3 R D particulate filters for the 8000 Series half mask. For protection against harmful and carcinogenic dusts, fumes and aerosols including radioactive particulates as well as airborne biological substances and enzymes. 20 x WEL.
Specifications
Specifications
Filter Type | P2RD |
Product Type | Particulate Filter |
For Use With (Equipment) | For Series 8000 Respiratory Filter |
Quantity | 8/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Moldex™ 8000 Particulate Filters >
Spot an opportunity for improvement?Share a Content Correction