Learn More
Uvex 95790 Insole For Uvex 1 Sport - Width 14
Comfortable climatic insole with antistatic properties for Uvex 1 Sport
Specifications
Color | Black |
---|---|
For Use With (Equipment) | Uvex 1 Sport |
Material | Microvelour |
Disposable | Reusable |
Gender | Unisex |
Description
- Full-surface foot shock absorption
- Highly breathable and moisture-absorbent material
- Shock absorption zones support natural foot movement
- Skin compatibility and pleasant surface structure
- Supports foot arch
- Article No. 9579.0
- Sizes: 35 to 52
Specifications
Black | |
Microvelour | |
Unisex | |
Yes |
Uvex 1 Sport | |
Reusable | |
Wide Width | |
No |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Uvex 95790 Insole For Uvex 1 Sport - Width 14