Learn More
Uvex™ 9770 Pheos ES Unventilated Safety Helmets
Lightweight, unventilated helmets in a sporty design ideal for use in the electrician sector with low voltage systems. Features a shortened shield with a conventional interior for easy adjustment.
Brand:  Uvex™ 9770321
Additional Details : Weight : 0.39800kg
 View more versions of this product
Product Code. 17269101
  • 150.00 SEK / Each
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
Description
Description
  • Lightweight, unventilated helmet in a sporty design
  • Completely closed for use in the electrician sector with low voltage systems
  • Includes an adapter system for visors, helmet caps, and helmet lamps
  • Conventional interior for easy adjustment
  • Shortened shield
Specifications
Specifications
EN 397:2012, EN 50365:2002
Low Voltage Systems
HDPE
Electrically Insulating Helmet
Not Ventilated
Red
Adapter System for Uvex™ Visors, Helmet Capsules, and Helmet Lamps
9770 Pheos ES
52 to 61 cm