missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20681-1-AP-20UL
This item is not returnable.
View return policy
Description
Probable transporter, its natural substrate has not been found yet. May have a role in macrophage lipid metabolism and neural development.Specifications
ABCA2 | |
Polyclonal | |
Unconjugated | |
ABCA2 | |
ABC transporter ABCA2, ABC2, ABCA2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11305, 20, 79248 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P41234, Q9BZC7, Q9ESR9 | |
Abca2 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ABCA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction