missing translation for 'onlineSavingsMsg'
Learn More

ABCA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20681-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16880255

  • 4721.67 SEK / 150µL
Estimated Shipment: 20-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Probable transporter, its natural substrate has not been found yet. May have a role in macrophage lipid metabolism and neural development.
Specifications

Specifications

ABCA2
Polyclonal
Unconjugated
ABCA2
ABC transporter ABCA2, ABC2, ABCA2
Rabbit
Antigen Affinity Chromatography
RUO
11305, 20, 79248
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P41234, Q9BZC7, Q9ESR9
Abca2
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers