Learn More
ABHD6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20494-1-AP-150UL
This item is not returnable.
View return policy
Description
ABHD6 has 2-arachidonoylglycerol hydrolase activity. ABHD6 may be a regulator of endocannabinoid signaling pathways.Specifications
ABHD6 | |
Polyclonal | |
Unconjugated | |
ABHD6 | |
ABHD6, Monoacylglycerol lipase ABHD6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57406, 66082 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8R2Y0, Q9BV23 | |
Abhd6 | |
ABHD6 Fusion Protein Ag13968 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Product Title
ABHD6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction