missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11111-1-AP-20UL
This item is not returnable.
View return policy
Description
ACAA2 (3-Ketoacyl-CoA thiolase) is an enzyme that in humans is encoded by the ACAA2 gene. Acetyl-Coenzyme A acyltransferase 2 is an acetyl-CoA C-acyltransferase enzyme. The encoded protein catalyzes the last step of the mitochondrial fatty acid beta-oxidation spiral. Unlike most mitochondrial matrix proteins, it contains a non-cleavable amino-terminal targeting signal.Specifications
ACAA2 | |
Polyclonal | |
Unconjugated | |
ACAA2 | |
ACAA2, Acetyl CoA acyltransferase, Beta ketothiolase, DSAEC, T1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10449, 170465, 52538 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P13437, P42765, Q8BWT1 | |
Acaa2 | |
ACAA2 Fusion Protein Ag1609 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ACAA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction