Learn More
ACADS Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16623-1-AP-20UL
This item is not returnable.
View return policy
Description
Introduces a double bond at position 2 in saturated acyl-CoA′s of short chain length, i.e. less than 6 carbon atoms.Specifications
ACADS | |
Polyclonal | |
Unconjugated | |
ACADS | |
ACAD3, ACADS, Butyryl CoA dehydrogenase, SCAD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11409, 35, 64304 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P15651, P16219, Q07417 | |
ACADS | |
ACADS Fusion Protein Ag9922 | |
20 ÎĽL | |
Primary | |
Non-human Primate, Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Product Title
ACADS Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech > 20 ÎĽL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction