Learn More
ACBD5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21080-1-AP-150UL
 View more versions of this product
Product Code. 16880685
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
This gene encodes a member of the acyl-Coenzyme A binding protein family, known to function in the transport and distribution of long chain acyl-Coenzyme A in cells. This gene may play a role in the differentiation of megakaryocytes and formation of platelets. A related protein in yeast is involved in autophagy of peroxisomes. A mutation in this gene has been associated with autosomal domit thrombocytopenia. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
ACBD5
Polyclonal
Unconjugated
ACBD5
ACBD5, KIAA1996
Rabbit
Antigen Affinity Chromatography
RUO
74159, 91452
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5T8D3, Q5XG73
Acbd5
ACBD5 Fusion Protein Ag15287
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ACBD5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated