Learn More
Acetate Buffer Solution, Fisher Chemical™ Product Code.: 12930684
Acetate Buffer Solution, Fisher Chemical™
BIOPHARMA
Product Code. 12930684 Shop All Fisher Chemical Products
1000 mL, 1000mL
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
Product Code. 12930684
Brand: Fisher Chemical SOL 039 1L
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Quantity 1000 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number2506003 Certificate TypeCertificate of Analysis Date03/06/2025 Product CodeSOL 039 1L
Lot Number1093659 Certificate TypeCertificate of Analysis Date30/03/2025 Product CodeSOL 039 1L
Lot Number1004159 Certificate TypeCertificate of Analysis Date04/03/2025 Product CodeSOL 039 1L
Lot Number1013913 Certificate TypeCertificate of Analysis Date04/03/2025 Product CodeSOL 039 1L
Lot Number1048339 Certificate TypeCertificate of Analysis Date03/03/2025 Product CodeSOL 039 1L
Product Content Correction
Product Title
Acetate Buffer Solution, Fisher Chemical™ >