missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOXL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23366-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
ACOXL gene ontology annotations related to this gene include fatty acid beta-oxidation.Specifications
ACOXL | |
Polyclonal | |
Unconjugated | |
ACOXL | |
ACOXL, Acyl CoA oxidase like protein, acyl Coenzyme A oxidase like | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55289 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NUZ1 | |
ACOXL | |
ACOXL Fusion Protein Ag18625 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |