missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOXL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | ACOXL |
---|---|
Concentration | 0.45 mg/mL |
Applications | Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16882315
|
Proteintech
23366-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16872315
|
Proteintech
23366-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACOXL gene ontology annotations related to this gene include fatty acid beta-oxidation.Specifications
ACOXL | |
Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q9NUZ1 | |
55289 | |
ACOXL Fusion Protein Ag18625 | |
Primary | |
-20°C | |
ACOXL |
0.45 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ACOXL, Acyl CoA oxidase like protein, acyl Coenzyme A oxidase like | |
ACOXL | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ACOXL Rabbit anti-Human, Polyclonal, Proteintech