missing translation for 'onlineSavingsMsg'
Learn More

ACPT Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 24326-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16882805

  • 1355.00 SEK / 20µL
Estimated Shipment: 31-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Acid phosphatases are enzymes capable of hydrolyzing orthophosphoric acid esters in an acid medium. This gene is up-regulated by androgens and is down-regulated by estrogens in the prostate cancer cell line. This gene exhibits a lower level of expression in testicular cancer tissues than in normal tissues. The protein encoded by this gene has structural similarity to prostatic and lysosomal acid phosphatases. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications

Specifications

ACPT
Polyclonal
Unconjugated
ACP4
acid phosphatase, testicular, ACPT, Testicular acid phosphatase
Rabbit
Antigen Affinity Chromatography
RUO
100503991, 93650
-20°C
Liquid
Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BZG2
Acp4
ACPT Fusion Protein Ag19283
20 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers