Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters for Mass Spectrometry
Acrodisc MS syringe filters are designed specifically to extend the lifespan of HPLC and UHPLC columns used in LC/MS analytical sample preparation. The use of syringe filters also reduces the amount of particulate entering the instrumentation, thus reducing chances of breakdown or failure.
Brand: Cytiva (formerly Pall Lab) MS-3301
Description
- Available in 13 and 25 mm diameters with 0.2 ÎĽm rated wwPTFE membrane and HDPE housings
- Constructed with a water-wettable polytetrafluoroethylene (wwPTFE) membrane in high density polyethylene (HDPE) housings
- The efficiency of the filters at removing particulates was examined using 0.05% polymer microsphere suspensions with an average diameter of 0.31 ÎĽm
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
wwPTFE | |
Non-sterile | |
HDPE |
0.2 ÎĽm | |
1 cm2 | |
13 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.