Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters for Mass Spectrometry
Acrodisc MS syringe filters are designed specifically to extend the lifespan of HPLC and UHPLC columns used in LC/MS analytical sample preparation. The use of syringe filters also reduces the amount of particulate entering the instrumentation, thus reducing chances of breakdown or failure.
Brand:  Cytiva (formerly Pall Lab) MS-3301
 View more versions of this product
Product Code. 17114761
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Available in 13 and 25 mm diameters with 0.2 ÎĽm rated wwPTFE membrane and HDPE housings
  • Constructed with a water-wettable polytetrafluoroethylene (wwPTFE) membrane in high density polyethylene (HDPE) housings
  • The efficiency of the filters at removing particulates was examined using 0.05% polymer microsphere suspensions with an average diameter of 0.31 ÎĽm
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
wwPTFE
Non-sterile
HDPE
0.2 ÎĽm
1 cm2
13 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters for Mass Spectrometry > 13 mm