missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACRV1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14040-1-AP-150UL
This item is not returnable.
View return policy
Description
One of the most frequent causes of man infertility is defective sperm acrosome. This damage can be detected using antibodies against intra-acrosomal proteins. Besides diagnostics of sperm pathology, monoclonal antibodies against intra-acrosomal proteins can be used for evaluation of the physiological state of sperm cells as well as for selection of a suitable method of fertilization in the laboratories of assisted reproduction.Specifications
ACRV1 | |
Polyclonal | |
Unconjugated | |
ACRV1 | |
Acrosomal protein SP 10, acrosomal vesicle protein 1, ACRV1, D11S4365, SP 10, SPACA2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P26436 | |
ACRV1 | |
ACRV1 Fusion Protein Ag5109 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ACRV1 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction