missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSBG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16077-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene possesses long-chain acyl-CoA synthetase activity. It is thought to play a central role in brain very long-chain fatty acids metabolism and myelinogenesis.Specifications
ACSBG1 | |
Polyclonal | |
Unconjugated | |
ACSBG1 | |
ACSBG1, BG1, BGM, GR LACS, hBG1, hsBG, hsBGM, KIAA0631, Lipodisin, LPD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171410, 23205, 94180 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q924N5, Q96GR2, Q99PU5 | |
Acsbg1 | |
ACSBG1 Fusion Protein Ag9043 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ACSBG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction