missing translation for 'onlineSavingsMsg'
Learn More

ACSM2A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 22862-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16882115

  • 1355.00 SEK / 20µL
Estimated Shipment: 31-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a mitochondrial acyl-coenzyme A synthetase that is specific for medium chain fatty acids. These enzymes catalyze fatty acid activation, the first step of fatty acid metabolism, through the transfer of acyl-CoA. These enzymes also participate in the glycine conjugation pathway in the detoxification of xenobiotics such as benzoate and ibuprofen. Expression levels of this gene in the kidney may be correlated with kidney function. This gene and its paralog ACSM2B (Gene ID: 348158), both present on chromosome 16, likely arose from a chromosomal duplication event.
Specifications

Specifications

ACSM2A
Polyclonal
Unconjugated
ACSM2A
A 923A4.1, ACSM2, ACSM2A, Butyrate CoA ligase 2A, MACS2
Rabbit
Antigen Affinity Chromatography
RUO
123876
-20°C
Liquid
Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q08AH3
ACSM2A
ACSM2A Fusion Protein Ag18856
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers