missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals Activin C/Inhibin beta C Recombinant Protein Antigen

Brand:  Novus Biologicals NBP2-76513PEP

Product Code. 18666561

  • 3055.00 SEK / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Antibody Competition
3626
0.1 mL
inhibin beta C chain, IHBC, inhibin, beta C
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-76513It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL.For current lot information, including availability, please contact our technical support team click nb-technical@bio-techne.com For further blocking peptide related information and a protocol, click here .
>80% by SDS-PAGE and Coomassie blue staining
PBS and 1M Urea, pH 7.4.
M.W. (Theoretical): 26 kDa
Store at -20C. Avoid freeze-thaw cycles.
INHBC
Cytokine Research
Activin C/Inhibin beta C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals Activin C/Inhibin beta C Recombinant Protein Antigen > 0.1 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.