missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTRT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16992-1-AP-150UL
This item is not returnable.
View return policy
Specifications
ACTRT2 | |
Polyclonal | |
Unconjugated | |
ACTRT2 | |
Actin related protein M2, actin related protein T2, ACTRT2, Arp T2, ARPM2, ARPT2, HARPM2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140625, 298671, 73353 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8TDY3, Q9D9L5 | |
Actrt2 | |
ACTRT2 Fusion Protein Ag10597 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ACTRT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction