Learn More
Socorex™ Acura Variable Volume Manual Pipettes
Secure the stability of your settings when pipetting with Socorex™ Acura Variable Volume Manual Pipettes. Each individual unit's accuracy has been measured for reliability at 0.5μL and 1μL.
Brand: Socorex™ 825.0010
2820.49 SEK valid until 2025-07-31
Use promo code "24075" to get your promotional price.
Description
- Constructed of polypropylene
- With precision setting mechanism
- Single-handed volume setting
Specifications
Autoclavable | |
835 | |
Acura | |
0.5 to 10 μL |
Polypropylene | |
1 | |
Micropipette | |
1 Each |
Promotions
-
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-07-31