Learn More
WTW™ ADA-DIN-BNC Model Adapter
Product Code. 10587451
Change view
Click to view available options
Connector Types:
BNC Plug, DIN Socket
Unit Size:
Each
Product Code. Connector Types Unit Size
10587451 BNC Plug, DIN Socket Each
1 options
Product Code. 10587451 Supplier WTW™ Supplier No. 108509
  • ONLINE EXCLUSIVE PRICE
    1085.00 SEK
    945.87 SEK / Paket med 1
    Save 139.13 SEK
    12% Av
View all promotions
I lager
Specifications
Connector Types BNC Plug, DIN Socket
For Use With (Equipment) pH Electrode
Type Adapter
For Use With (Application) Connecting pH Electrodes with BNC Plug to a Meter with DIN Socket