Learn More
ADA Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13328-1-AP-20UL
 View more versions of this product
Product Code. 16881474
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
Catalyzes the hydrolytic deamination of adenosine and 2-deoxyadenosine. Plays an important role in purine metabolism and in adenosine homeostasis. Modulates signaling by extracellular adenosine, and so contributes indirectly to cellular signaling events. May act as a positive regulator of T-cell coactivation.
Specifications
Specifications
ADA
Polyclonal
Unconjugated
ADA
ADA, Adenosine aminohydrolase, adenosine deaminase
Rabbit
Antigen Affinity Chromatography
RUO
100, 11486
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P00813, P03958
ADA
ADA Fusion Protein Ag4128
20 μL
Primary
Mouse, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ADA Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated