Learn More
Hettich™ Adapter for Carrier no. 1427
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Marque:  Hettich™ 5275
1015.70 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 Afficher plus de versions de ce produit
Code produit. 12043924
  • 1095.00 SEK / Each
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
  • Adapter for Carrier no. 1427
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Spécification
Spécification
Rotor Adapter
15 mL
120 mm
1
Conical
17 mm
Carrier no. 1427
1
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Product Title
Hettich™ Adapter for Carrier no. 1427 > 15mL