Learn More
Hettich™ Adapter for Carriers no. 4254, 4547-B, 4579-A, 4890
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 4434
Additional Details : Weight : 0.99000kg
 View more versions of this product
Product Code. 10783087
  • 1555.00 SEK / Each
Estimated Shipment: 08-07-2024
to see stock.
Description
Description
  • Adapter for Carriers no. 4254, 4547-B, 4579-A, 4890
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
Rotor Adapter
19 x 4 to 15 mL Tubes
102 mm
19
Blood/Urine, Round Bottom
17 mm
Carriers no. 4254, 4547-B, 4579-A, 4890