Learn More
Hettich™ Adapter for Carriers no. 4254, 4547-B, 4579-A, 4890
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 4438
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12033914
  • 2185.00 SEK / Each
Estimated Shipment: 17-07-2024
to see stock.
Description
Description
  • Adapter for Carriers no. 4254, 4547-B, 4579-A, 4890
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
Rotor Adapter
7 x 25 to 30 mL
110 mm
7
Conical, Skirted
25 mm
Carriers no. 4254, 4547-B, 4579-A, 4890