Learn More
Eppendorf™ Adapter for Rotors S-4-104, S-4x750, S-4x1000
Facilitates use of 30 mL conical tube array in rotor buckets for S-4-104, S-4x750, S-4x1000 rotors.
Brand:  Eppendorf™ 5825755006
3900.00 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15523584
  • 3915.60 SEK / Each
Description
  • Suitable for S-4-104, S-4x750, S-4x1000 rotor buckets
  • Accepts 8 x 30 mL conical tubes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Conical
S-4-104, S-4x750, S-4x1000 rotors and buckets
8
8 x 30 mL Tubes
8
2/Pk.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Eppendorf™ Adapter for Rotors S-4-104, S-4x750, S-4x1000 > 8 x 30 mL tubes