Learn More
Fisherbrand™ Adapters for Fisherbrand™ accuSpin™ 400 6-Place Fixed-Angle Rotor
Brand:  Fisherbrand™ 75003095
987.62 SEK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Product Code. 10658612
  • 1042.45 SEK / Each
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Description
For use with fixed-angle rotor 13-100-559.
Adapters are sold individually. Order the correct quantity to outfit rotor.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Conical
400 6-place Fixed angle rotor
15 mL
SDS
Documents Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Save up to 59% on some of our most popular products
    Promo Code: 24140
    Expires2025-08-24
    Days
    57
    Hours
    8
    Minutes
    27
    Seconds
    52
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Days
    2
    Hours
    8
    Minutes
    27
    Seconds
    52
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
  • Multiple Manufacturers
    Get amazing savings on products for your analyses
    Promo Code: 24135
    Expires2025-07-13
    Days
    16
    Hours
    8
    Minutes
    27
    Seconds
    52
  • Multiple Manufacturers
    Browse exclusive prices for our academic customers
    Promo Code: 25081
    Expires2025-12-31
  • Multiple Manufacturers
    Save big on essential products for cancer research
    Promo Code24147
    Expires2025-07-31
Product Content Correction
Product Title
Fisherbrand™ Adapters for Fisherbrand™ accuSpin™ 400 6-Place Fixed-Angle Rotor > 15mL conical