Learn More
3M™ Adflo™ Gas Filter, A2, 837542
Click to view available options
:
Category: Welding Parts and Accessories, Model: 83 75 42
Description
- Provides protection against a range of particulate and gas hazards (dependent upon filters selected)
- For use with 3M™ Adflo™ Powered Air Purifying Respirators (PAPR)
- Particle filter, gas filters, and odour filter/pad available
- Installation is easy and secure
- For highest possible safety and comfort, regularly check your equipment for worn or damaged parts
Specifications
Specifications
Cartridge Type | Organic Vapor, Acid Gas |
Color | Black |
Product Type | Filter |
Product Content Correction
Product Title
3M™ Adflo™ Gas Filter, A2, 837542 >
Spot an opportunity for improvement?Share a Content Correction