Learn More
ADH1B Mouse anti-Human, Unconjugated, Clone: 1F1B4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66939-1-IG-20UL
Description
The ADH1B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ADH1B. This antibody recognizes human, mouse, rat, pig antigen. The ADH1B antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
ADH1B | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, Immunohistochemistry, Immunofluorescence | |
1F1B4 | |
Unconjugated | |
20 μL | |
125 | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.