Learn More
ADH1B Mouse anti-Human, Unconjugated, Clone: 1F1B4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66939-1-IG-150UL
 View more versions of this product
Product Code. 16879245
  • 4652.90 SEK / 150µL
Description
The ADH1B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ADH1B. This antibody recognizes human, mouse, rat, pig antigen. The ADH1B antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
ADH1B
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, Immunofluorescence
1F1B4
Unconjugated
150 μL
125
IgG2a
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
ADH1B Mouse anti-Human, Unconjugated, Clone: 1F1B4, Proteintech > 150 μL