missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADPGK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | ADPGK |
---|---|
Concentration | 0.15 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16894894
|
Proteintech
15639-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16884894
|
Proteintech
15639-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ADPGK.Specifications
ADPGK | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q8VDL4, Q9BRR6 | |
315722, 72141, 83440 | |
ADPGK Fusion Protein Ag8105 | |
Primary | |
-20°C | |
ADPGK |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
2610017G09Rik, ADP dependent glucokinase, ADP GK, ADPGK, RbBP 35 | |
Adpgk | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ADPGK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech