Learn More
ADRA2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19778-1-AP-20UL
 View more versions of this product
Product Code. 16879254
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The antibody is specific to ADRA2B.
Alpha-2-adrenergic receptors are members of the G protein-coupled receptor superfamily. They include 3 highly homologous subtypes: alpha2A, alpha2B, and alpha2C. These receptors have a critical role in regulating neurotransmitter release from sympathetic nerves and from adrenergic neurons in the central nervous system. Alpha 2B adrenergic receptor subtype was observed to associate with eIF-2B, a guanine nucleotide exchange protein that functions in regulation of translation. A polymorphic variant of the alpha2B subtype, which lacks 3 glutamic acids from a glutamic acid repeat element, was identified to have decreased G protein-coupled receptor kinase-mediated phosphorylation and desensitization; this polymorphic form is also associated with reduced basal metabolic rate in obese subjects and may therefore contribute to the pathogenesis of obesity. Alpha 2B adrenergic receptor gene contains no introns in either its coding or untranslated sequences.
Specifications
Specifications
ADRA2B
Polyclonal
Unconjugated
ADRA2B
ADRA2B, ADRA2L1, ADRA2RL1, ADRARL1, Alpha 2B adrenergic receptor, Alpha 2B adrenoceptor, Alpha 2B adrenoreceptor, Alpha 2BAR, ALPHA2BAR
Rabbit
Antigen Affinity Chromatography
RUO
11552, 151, 24174
-20°C
Liquid
Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin)
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P18089, P19328, P30545
Adra2b
Peptide
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ADRA2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated