missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AEBP2 Mouse anti-Human, Porcine, Clone: 7H2B11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66141-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The antibody recognizes adult-specific larger form (52 kDa) and the embryo-specific smaller form (31 kDa).
AEBP2 is a DNA-binding transcriptional repressor. It may interact with and stimulate the activity of the PRC2 complex, which methylates ′Lys-9′ and ′Lys-27′ residues of histone H3.Specifications
AEBP2 | |
Monoclonal | |
0.93 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6ZN18 | |
AEBP2 | |
AEBP2 Fusion Protein Ag17905 | |
20 μL | |
Primary | |
Human, Porcine | |
Antibody | |
IgG1 |
Western Blot | |
7H2B11 | |
Unconjugated | |
AEBP2 | |
AE binding protein 2, AEBP2, Zinc finger protein AEBP2 | |
Mouse | |
Protein G | |
RUO | |
100522805, 121536 | |
-20°C | |
Liquid |