missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AF-6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1662.46 SEK - 4700.76 SEK
Specifications
Antigen | AF-6 |
---|---|
Concentration | 0.82 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16896545
|
Proteintech
55102-1-AP-20UL |
20 ÎĽL |
1662.46 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16886545
|
Proteintech
55102-1-AP-150UL |
150 ÎĽL |
4700.76 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody is specific to MLLT4.
AFDN belongs to an adhesion system, probably together with the E-cadherin-catenin system, which plays a role in the organization of homotypic, interneuronal and heterotypic cell-cell adherens junctions (AJs). Nectin- and actin-filament-binding protein that connects nectin to the actin cytoskeleton.Specifications
AF-6 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
O35889, P55196, Q9QZQ1 | |
17356, 26955, 4301 | |
Peptide | |
Primary | |
-20°C | |
AFDN |
0.82 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AF 6, AF6, AFADIN, Afadin variant, MLLT4 | |
Afdn | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AF-6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech