missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGFG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12670-1-AP-150UL
This item is not returnable.
View return policy
Description
The ADP-ribosylation factor (Arf) family are highly conserved members of the Ras superfamily of regulatory GTP-binding proteins. Arf proteins participate in routing of intracellular proteins to and within the Golgi complex. Cellular functions include maintece of organelle integrity, coat protein assembly, as an activator of phospholipase D. The Arf family is divided functionally into the Arf and the Arf-like (Arl) proteins. The ARF proteins are categorized as class I (ARF1, ARF2,and ARF3), class II (ARF4 and ARF5) and class III (ARF6) and members of each class share a common gene organization.Specifications
AGFG1 | |
Polyclonal | |
Unconjugated | |
AGFG1 | |
AGFG1, ArfGAP with FG repeats 1, HIV 1 Rev binding protein, HRB, Nucleoporin like protein RIP, RAB, Rev interacting protein, RIP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15463, 3267, 363266 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P52594, Q4KLH5, Q8K2K6 | |
AGFG1 | |
AGFG1 Fusion Protein Ag3352 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AGFG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction