missing translation for 'onlineSavingsMsg'
Learn More

AGR2 Mouse anti-Human, Porcine, Clone: 1A8A8, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66768-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16888945

  • 1355.00 SEK / 20µL
Estimated Shipment: 03-04-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

AGR2 (anterior gradient protein 2) and AGR3 are members of the anterior gradient homolog family. They are secreted cytoplasmic proteins which are involved in metastasis induction and p53 tumour supressor inhibition. They may serve as molecular markers and potential therapeutic targets for hormone-responsive breast tumours. AGR2 is ubiquitously expressed with upregulated expression in prostate cancer, breast cancer, lung cancer, renal carcinomas and endometrial carcinomas. AGR2 expression is positively correlated with that of the estrogen receptor (ER) and is negatively correlated with that of the epidermal growth factor receptor (EGFR).
Specifications

Specifications

AGR2
Monoclonal
0.93 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95994
AGR2
AGR2 Fusion Protein Ag2919
20 μL
Primary
Porcine, Human
Antibody
IgG2b
Western Blot
1A8A8
Unconjugated
AGR2
AG 2, AG2, AGR2, GOB 4, HAG 2, HPC8, PDIA17, XAG 2
Mouse
Protein A
RUO
100514482, 10551
-20°C
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers