missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGTR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25343-1-AP-20UL
This item is not returnable.
View return policy
Description
AGTR1 is known as Angiotensin II Type 1 Receptor. Angiotensin II is a potent vasopressor hormone and a primary regulator of aldosterone secretion. It is an important effector controlling blood pressure and volume in the cardiovascular system. And it acts through at least two types of receptors. This gene encodes the type 1 receptor which is thought to mediate the major cardiovascular effects of angiotensin II. Moreover, this gene may play a role in the generation of reperfusion arrhythmias following restoration of blood flow to ischemic or infarcted myocardium. It was previously thought that a related gene, denoted as AGTR1B, existed; however, it is now believed that there is only one type 1 receptor gene in humans. Multiple alternatively spliced transcript variants have been reported for this gene.Specifications
AGTR1 | |
Polyclonal | |
Unconjugated | |
AGTR1 | |
AG2S, AGTR1, AGTR1A, AGTR1B, Angiotensin II type 1 receptor, AT1, AT1AR, AT1B, AT1BR, AT1R, AT2R1, AT2R1A, AT2R1B, HAT1R, Type 1 angiotensin II receptor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11607, 185, 24180 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P25095, P29754, P30556 | |
AGTR1, Agtr1a | |
AGTR1 Fusion Protein Ag14461 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AGTR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction