Learn More
AHCYL2 Mouse anti-Human, Unconjugated, Clone: 1D1F6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67484-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16870136
  • 4721.67 SEK / 150µL
Estimated Shipment: 14-05-2024
to see stock.
Description
Description
The AHCYL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AHCYL2. This antibody recognizes human antigen. The AHCYL2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
AHCYL2
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1D1F6
Unconjugated
150 μL
23382
IgG2a