Learn More
AHCYL2 Mouse anti-Human, Unconjugated, Clone: 1D1F6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67484-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The AHCYL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AHCYL2. This antibody recognizes human antigen. The AHCYL2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
AHCYL2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
1D1F6 | |
Unconjugated | |
150 μL | |
23382 | |
IgG2a |