Learn More
AHCYL2 Mouse anti-Human, Unconjugated, Clone: 1D1F6, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | AHCYL2 |
---|---|
Clone | 1D1F6 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16880136
|
Proteintech
67484-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16870136
|
Proteintech
67484-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The AHCYL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AHCYL2. This antibody recognizes human antigen. The AHCYL2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
AHCYL2 | |
1000 ÎĽg/mL | |
Monoclonal | |
Mouse | |
23382 | |
Primary |
1D1F6 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Human | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.