missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20590-1-AP-20UL
This item is not returnable.
View return policy
Description
AIM2 is a member of the IFI20X /IFI16 family. It plays a putative role in tumorigenic reversion and may control cell proliferation. Interferon-gamma induces expression of AIM2.Specifications
AIM2 | |
Polyclonal | |
Unconjugated | |
AIM2 | |
absent in melanoma 2, AIM2, PYHIN4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
304987, 383619, 9447 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.57 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14862, Q91VJ1 | |
Aim2 | |
AIM2 Fusion Protein Ag14520 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AIM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction