missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP12 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25199-1-AP-20UL
This item is not returnable.
View return policy
Description
The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins, which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. The encoded protein is expressed in endothelial cells, cultured fibroblasts, and osteosarcoma cells. It associates with protein kinases A and C and phosphatase, and serves as a scaffold protein in signal transduction. This protein and RII PKA colocalize at the cell periphery. This protein is a cell growth-related protein. Antibodies to this protein can be produced by patients with myasthenia gravis. Alternative splicing of this gene results in two transcript variants encoding different isoforms.Specifications
AKAP12 | |
Polyclonal | |
Unconjugated | |
AKAP12 | |
A kinase anchor protein 12, AKAP 12, AKAP 250, AKAP12, AKAP250, Gravin, Myasthenia gravis autoantigen | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
83425, 9590 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q02952, Q5QD51 | |
AKAP12 | |
AKAP12 Fusion Protein Ag17769 | |
20 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
AKAP12 Rabbit anti-Human, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction