missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR1C3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11194-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
DD1 is also designated AKR1C1, DDH or DDH1 while DD2 also can be designated AKR1C2, dDD, BABP or DDH2. AKR1C3 and 3alpha-HSD are alternate designations for DD3, while DD4 also can be called AKR1C4, CD or CHDR. DD3 is a unique enzyme that can specifically catalyze the dehydrogenation of trans-benzenedihydrodiol and trans-naphthalenedihydrodiol.Specifications
AKR1C3 | |
Polyclonal | |
Unconjugated | |
AKR1C3 | |
17 beta HSD 5, 3 alpha HSD type 2, 3 alpha HSD type II, brain, AKR1C3, DD 3, DD3, DDX, Dihydrodiol dehydrogenase 3, HA1753, HAKRB, HAKRe, hluPGFS, HSD17B5, Indanol dehydrogenase, KIAA0119, PGFS, Prostaglandin F synthase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8644 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P42330 | |
AKR1C3 | |
AKR1C3 Fusion Protein Ag1674 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |