missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR1C3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | AKR1C3 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828113
|
Proteintech
11194-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16818113
|
Proteintech
11194-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
DD1 is also designated AKR1C1, DDH or DDH1 while DD2 also can be designated AKR1C2, dDD, BABP or DDH2. AKR1C3 and 3alpha-HSD are alternate designations for DD3, while DD4 also can be called AKR1C4, CD or CHDR. DD3 is a unique enzyme that can specifically catalyze the dehydrogenation of trans-benzenedihydrodiol and trans-naphthalenedihydrodiol.Specifications
AKR1C3 | |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
P42330 | |
8644 | |
AKR1C3 Fusion Protein Ag1674 | |
Primary | |
-20°C | |
AKR1C3 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
17 beta HSD 5, 3 alpha HSD type 2, 3 alpha HSD type II, brain, AKR1C3, DD 3, DD3, DDX, Dihydrodiol dehydrogenase 3, HA1753, HAKRB, HAKRe, hluPGFS, HSD17B5, Indanol dehydrogenase, KIAA0119, PGFS, Prostaglandin F synthase | |
AKR1C3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |