missing translation for 'onlineSavingsMsg'
Learn More

AKR1C3 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11194-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16828113

  • 1656.73 SEK / 20µL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

DD1 is also designated AKR1C1, DDH or DDH1 while DD2 also can be designated AKR1C2, dDD, BABP or DDH2. AKR1C3 and 3alpha-HSD are alternate designations for DD3, while DD4 also can be called AKR1C4, CD or CHDR. DD3 is a unique enzyme that can specifically catalyze the dehydrogenation of trans-benzenedihydrodiol and trans-naphthalenedihydrodiol.
Specifications

Specifications

AKR1C3
Polyclonal
Unconjugated
AKR1C3
17 beta HSD 5, 3 alpha HSD type 2, 3 alpha HSD type II, brain, AKR1C3, DD 3, DD3, DDX, Dihydrodiol dehydrogenase 3, HA1753, HAKRB, HAKRe, hluPGFS, HSD17B5, Indanol dehydrogenase, KIAA0119, PGFS, Prostaglandin F synthase
Rabbit
Antigen Affinity Chromatography
RUO
8644
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P42330
AKR1C3
AKR1C3 Fusion Protein Ag1674
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers