Learn More
Al-23 Insulating Tube, Both Ends Open (Thin Wall);OD (mm), 0.5;ID (mm), 0.2 Product Code.: 11307478
Al-23 Insulating Tube, Both Ends Open (Thin Wall);OD (mm), 0.5;ID (mm), 0.2
Product Code. 11307478
300 mm, 300mm
Click to view available options
Quantity:
100 mm
200 mm
250 mm
300 mm
Unit Size:
100mm
200mm
250mm
300mm
Product Code. 11307478
Brand: Thermo Scientific Alfa Aesar 031475.DW
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Insulating Tube
Name Note Both ends open (Thin Wall)
Quantity 300 mm
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberD14U008 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number031475.DW,031475.DT,030357.04,030357.02
Lot NumberE20Q15 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number031475.DW,031475.DT,030357.04,030357.02
Lot NumberY18F051 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number031475.DW,031475.DT,030357.04,030357.02
Lot NumberG07T029 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number031475.DW,031475.DT,030357.04,030357.02
Lot NumberU02E006 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number031475.DW,031475.DT,030357.04,030357.02
RUO – Research Use Only
Product Content Correction
Product Title
Al-23 Insulating Tube, Both Ends Open (Thin Wall);OD (mm), 0.5;ID (mm), 0.2 >