Learn More
Al-23 Multibore Insulating Tube, O. D.: 8mm, Two 1.6mm Bores, Thermo Scientific™ Product Code.: 11338447
Al-23 Multibore Insulating Tube, O. D.: 8mm, Two 1.6mm Bores, Thermo Scientific™
Product Code. 11338447
770 mm, 770mm
Click to view available options
Quantity:
205 mm
275 mm
380 mm
560 mm
770 mm
1060 mm
1460 mm
2060 mm
Unit Size:
1060mm
1460mm
205mm
2060mm
275mm
380mm
560mm
770mm
Product Code. 11338447
Brand: Thermo Scientific Alfa Aesar 032558.EL
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Multibore Insulating Tube
Name Note Two bores
Quantity 770 mm
Product Content Correction
Product Title
Al-23 Multibore Insulating Tube, O. D.: 8mm, Two 1.6mm Bores, Thermo Scientific™ >