Learn More
Al-23 Tube, Both Ends Open, O.D.: 3mm, I.D.: 1.6mm, Thermo Scientific™
236.53 SEK - 1264.21 SEK
Specifications
Chemical Name or Material Al-23 Tube
Name Note Both ends open
View More Specs
Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11363907
View Documents
Thermo Scientific Alfa Aesar
031553.DM
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11303917
View Documents
Thermo Scientific Alfa Aesar
031553.DR
200 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11373907
View Documents
Thermo Scientific Alfa Aesar
031553.DW
300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11353907
View Documents
Thermo Scientific Alfa Aesar
031553.EH
600 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11313917
View Documents
Thermo Scientific Alfa Aesar
031553.EM
800 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11393907
View Documents
Thermo Scientific Alfa Aesar
031553.EO
1000 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Al-23 Tube
Both ends open
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Al-23 Tube, Both Ends Open, O.D.: 3mm, I.D.: 1.6mm, Thermo Scientific™