Learn More
ALADIN Rabbit anti-Human, Non-human primate, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15127-1-AP-20UL
 View more versions of this product
Product Code. 16894164
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The protein encoded by this gene is a member of the WD-repeat family of regulatory proteins and may be involved in normal development of the peripheral and central nervous system. The encoded protein is part of the nuclear pore complex and is anchored there by NDC1. Defects in this gene are a cause of achalasia-addisonianism-alacrima syndrome (AAAS), also called triple-A syndrome or Allgrove syndrome. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
ALADIN
Polyclonal
Unconjugated
AAAS
AAA, AAAS, AAASb, ADRACALA, ADRACALIN, ALADIN, GL003
Rabbit
Antigen Affinity Chromatography
RUO
8086
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry
0.49 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NRG9
AAAS
ALADIN Fusion Protein Ag7211
20 ÎĽL
Primary
Human, Non-human Primate
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ALADIN Rabbit anti-Human, Non-human primate, Polyclonal, Proteintech > 20 ÎĽL; Unconjugated