Learn More
Aldosterone, TRC
Product Code. 30003653
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003653 5 mg 5mg
30004158 1 mg 1mg
30003398 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30003653 Supplier Toronto Research Chemicals Supplier No. TRCA5147005MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 52-39-1
Molecular Formula C21 H28 O5
Molecular Weight (g/mol) 360.44
Synonym Pregn-4-en-18-al, 11,21-dihydroxy-3,20-dioxo-, (11β)-, Aldosterone (8CI), (+)-Aldosterone, 11β,21-Dihydroxy-3,20-diketo-4-pregnen-18-al, 11β,21-Dihydroxy-3,20-diketopregn-4-ene-18-al, 11β,21-Dihydroxypregn-4-ene-3,18,20-trione, 18-Formyl-11β,21-dihydroxy-4-pregnene-3,20-dione, 18-Oxocorticosterone, Aldocorten, Aldocortene, Aldocortin, Electrocortin, Elektrocortin, NSC 73856, Reichstein X, d-Aldosterone
IUPAC Name (8S,9S,10R,11S,13R,14S,17S)-11-hydroxy-17-(2-hydroxyacetyl)-10-methyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthrene-13-carbaldehyde
SMILES C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@H](C(=O)CO)[C@]4(C[C@H](O)[C@H]23)C=O
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C21H28O5/c1-20-7-6-13(24)8-12(20)2-3-14-15-4-5-16(18(26)10-22)21(15,11-23)9-17(25)19(14)20/h8,11,14-17,19,22,25H,2-7,9-10H2,1H3/t14-,15-,16+,17-,19+,20-,21+/m0/s1
Formula Weight 360.1937
Chemical Name or Material Aldosterone