Learn More
Thermo Scientific Chemicals Sapphire substrate, 50.8mm dia x 0.332mm thick, EPI polished one side, C-axis, HEMCOR single crystal
Brand:  Thermo Scientific Chemicals 045018.KU
 View more versions of this product
Product Code. 15433925
  • 12315.00 SEK / Pack of 5
Description
Sapphire is used as an insulating substrate upon which to deposit silicon to make the integrated circuits, high-power radio-frequency (RF) applications such as those found in cellular telephones, public-safety band radios, and satellite communication systems, used in the semiconductor industry, blue light-emitting diodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sapphire Substrate, HEMCOR Single Crystal
50.8 mm
50.8mm dia. x 0.332 mm thick
EPI polished one side
5 Ea.
0.332 mm
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberH06S010 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number045018.KT,045019.KT,045018.KU
Lot NumberY15D025 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number045018.KT,045019.KT,045018.KU
Lot NumberG03Y023 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number045018.KT,045019.KT,045018.KU
Lot NumberP22E003 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number045018.KT,045019.KT,045018.KU
Lot NumberS07A033 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number045018.KT,045019.KT,045018.KU
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific Chemicals Sapphire substrate, 50.8mm dia x 0.332mm thick, EPI polished one side, C-axis, HEMCOR single crystal > 5each